n-Tetratetracontane ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

05440025MG
100282-210EA 333.46 USD
100282-210
n-Tetratetracontane ≥97%
n-Tetratetracontane

Store in a cool, dry place

Formula: H₃C(CH₂)₄₂CH₃
Melting Pt: 86.6 °C
MDL Number: MFCD00015268
CAS Number: 7098-22-8
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef28242e3ae7e267f723387a64a4032ed77cd0d5&vid=ef28242e3ae7e267f723387a64a4032ed77cd0d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR