H-D-Tyr(tBu)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

049821GM
100282-094EA 227.54 USD
100282-094
H-D-Tyr(tBu)-OH ≥99%
H-D-Tyr(tBu)-OH
Formula: C₁₃H₁₉NO₃
MW: 237.299 g/mol
MDL Number: MFCD00077112
CAS Number: 186698-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0241d07b8bc4104c76b70e3f1a35c42b4a9aea50&vid=15ea7938a5f8d0b08c6d4908e493c3cb02c713f2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR