Boc-D-Asn(Xan)-OH ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

049451GM
100282-054EA 189.62 USD
100282-054
Boc-D-Asn(Xan)-OH ≥99%, powder
Boc-D-Asn(Xan)-OH

  • White powder
  • Store at 0-8 °C

Formula: C₂₂H₂₄N₂O₆
MW: 412.442 g/mol
MDL Number: MFCD00076910
CAS Number: 200192-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR