(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine

049201GM
100282-012EA 149.08 USD
100282-012
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid ≥99%, powder
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid

  • White powder
  • Store at 0-8 °C

Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Melting Pt: 92…96 °C
MDL Number: MFCD00065603
CAS Number: 53267-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4716c9690ed75b7655f880c633940f654caffa9b&vid=40aa74aaa001162d6899e7d1327f8f1ed21c47b9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR