Boc-D-His(Tos)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

048951GM
100281-970EA 160.85 USD
100281-970
Boc-D-His(Tos)-OH ≥99%
Boc-D-His(Tos)-OH
Formula: C₁₈H₂₃N₃O₆S
MW: 409.5 g/mol
MDL Number: MFCD00037890
CAS Number: 69541-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d5c6ae26480bf319d4cf9685416cac2c5eaf41&vid=62253e7811e48e197fc6f273dd1ec55833d0cda9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR