Boc-PHG-OSu ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

048491GM
100281-894EA 151.69 USD
100281-894
Boc-PHG-OSu ≥99%
Boc-PHG-OSu
Formula: C₁₇H₂₀N₂O₆
MW: 348.355 g/mol
MDL Number: MFCD00190826
CAS Number: 201152-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=626da7d050450ebf8098f81c49147d2a48e01119&vid=626da7d050450ebf8098f81c49147d2a48e01119&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR