Boc-D-Phe-OSu ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-D-phenylalanine N-hydroxysuccinimide ester, (R)-2,5-Ddioxopyrrolidin-1-yl 2-((tert-butoxycarbonyl)amino)-3-phenylpropanoate

048481GM
100281-892EA 37.92 USD
100281-892
Boc-D-Phe-OSu ≥99%, powder
Boc-D-Phe-OSu

  • White powder
  • Store at 0-8 °C

Formula: C₁₈H₂₂N₂O₆
MW: 362.38 g/mol
Melting Pt: 166…173 °C
MDL Number: MFCD00270515
CAS Number: 3674-18-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c7db7d663deffa0d6b2339638329fc33666d0d&vid=09c7db7d663deffa0d6b2339638329fc33666d0d&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6750dcd01ba146e35c613bc8722147c2afb46d3&vid=c6750dcd01ba146e35c613bc8722147c2afb46d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR