Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nɑ-Boc-D-lysine, Boc-D-Lys-OH

047631GM
100281-770EA 85 USD
100281-770
Nɑ-(tert-Butoxycarbonyl)-D-lysine ≥98%, powder
Nalpha-(tert-Butoxycarbonyl)-D-lysine

  • White powder
  • Store at 0-8 °C

Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
Melting Pt: 192…212 °C
MDL Number: MFCD00076956
CAS Number: 106719-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee0f90773286142b95362241ddd82a807548c748&vid=380cd1f19f05e2d9b56785f04e027c537ae5bd12&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR