Boc-trans-4-hydroxy-L-proline ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline

047355GM
100281-718EA 41.85 USD
100281-718
Boc-trans-4-hydroxy-L-proline ≥99%, powder
Boc-trans-4-hydroxy-L-proline

  • White to off-white powder
  • Store at 0-8 °C

Formula: C₁₀H₁₇NO₅
MW: 231.3 g/mol
Melting Pt: 119…125 °C
MDL Number: MFCD00053370
CAS Number: 13726-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=142923556db0a471f95bd7195bf8c824732498ff&vid=142923556db0a471f95bd7195bf8c824732498ff&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70b9bc4458bac53d6bcea902016a322043c4f94&vid=f70b9bc4458bac53d6bcea902016a322043c4f94&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR