Boc-Glu(OBzl)-ONp ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

046805GM
100281-640EA 138.62 USD
100281-640
Boc-Glu(OBzl)-ONp ≥98%
Boc-Glu(OBzl)-ONp
Formula: C₂₃H₂₆N₂O₈
MW: 458.467 g/mol
MDL Number: MFCD00038124
CAS Number: 7536-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR