H-D-Asp(OBzl)-OH ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

043505GM
100281-206EA 300.77 USD
100281-206
H-D-Asp(OBzl)-OH ≥99%, powder
H-D-Asp(OBzl)-OH

  • White powder
  • Store at -0 °C

Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD00077100
CAS Number: 13188-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce0c0b7106d49292002a102179f51220abf5dcc2&vid=ce0c0b7106d49292002a102179f51220abf5dcc2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR