Z-ABU-OSu ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

042551GM
100280-844EA 209.23 USD
100280-844
Z-ABU-OSu ≥98%
Z-ABU-OSu
Formula: C₁₆H₁₈N₂O₆
MDL Number: MFCD00191058
CAS Number: 71447-81-9
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2273de89e79191a151491d2504a6e591fc860da&vid=b2273de89e79191a151491d2504a6e591fc860da&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR