(R)-Pyrrolidine-2-carboxamide hydrochloride ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: D-Prolinamide hydrochloride, H-D-Pro-NH₂·HCl

0423621GM
100280-588EA 138.62 USD
100280-588
(R)-Pyrrolidine-2-carboxamide hydrochloride ≥98%, powder
(R)-Pyrrolidine-2-carboxamide hydrochloride

  • White Powder
  • Store at 0-8 °C

Formula: C₅H₁₁ClN₂O
MW: 150.606 g/mol
Melting Pt: 176…182 °C
MDL Number: MFCD00237235
CAS Number: 50894-62-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd3f0db8a295afe067e864cb257c127b35db29d2&vid=381cfe15a98c90606da48fa3a00e0a7c21985cc5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR