D-(-)-Ornithine hydrochloride ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Orn-OH·HCl, H-D-Orn-OH.HCl

0422205GM
100280-384EA 122.92 USD
100280-384
D-(-)-Ornithine hydrochloride ≥99%, crystalline powder
D(-)-Ornithine hydrochloride

  • White to off-white crystalline powder
  • Store in a cool, dry place

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
Melting Pt: 239 °C (dec.)
MDL Number: MFCD00012917
CAS Number: 16682-12-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6812294ef0b0a37b9dbd3553b431b50202d5dac7&vid=6812294ef0b0a37b9dbd3553b431b50202d5dac7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR