l-leucylglycylglycine

Supplier: INDOFINE CHEMICAL COMPANY MS

0419555GM
100280-020EA 490.38 USD
100280-020
l-leucylglycylglycine
l-leucylglycylglycine
Formula: C₁₀H₁₉N₃O₄
MDL Number: MFCD00065942
CAS Number: 1187-50-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93a34f4d3bbd1154a1dc9c62de130fc08422a03c&vid=93a34f4d3bbd1154a1dc9c62de130fc08422a03c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR