2-Amino-4-methylpentanamide hydrochloride ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-DL-Leu-NH₂·HCl, H-DL-Leu-NH2.HCl

0418995GM
100279-928EA 98.08 USD
100279-928
2-Amino-4-methylpentanamide hydrochloride ≥98%, powder
2-Amino-4-methylpentanamide hydrochloride

  • White to off-white powder
  • Store at 0-8 °C

Formula: C₆H₁₅ClN₂O
Melting Pt: 222…228 °C
MDL Number: MFCD00070244
CAS Number: 10466-60-1
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32a151b5b0a8c3d19cbdd1e82dd5714431f694e0&vid=32a151b5b0a8c3d19cbdd1e82dd5714431f694e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR