H-D-Leu-NH₂·HCl ≥98%, crystals

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Leu-NH₂·HCl, H-D-Leu-NH2.HCl

0418981GM
100279-926EA 128.15 USD
100279-926
H-D-Leu-NH₂·HCl ≥98%, crystals
H-D-Leu-NH₂·HCl

  • Crystalline
  • Store at 0-8 °C

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD00083640
CAS Number: 80970-09-8
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c6cabbd36162a453cd74e3a859515d549f723d&vid=5352849898d802167726436f5a6ac4be000df188&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR