L-Hyp-(tBu)-OtBu·HCI ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Hyp(tBu)-OtBu.HCl, H-Hyp(tBu)-OtBu·HCl

0418261GM
100279-822EA 298.15 USD
100279-822
L-Hyp-(tBu)-OtBu·HCI ≥98%, powder
L-Hyp-(tBu)-OtBu·HCI

  • White powder
  • Store at 0-8 °C

Formula: C₁₃H₂₅NO₃·HCl
MW: 279.84 g/mol
Melting Pt: 140.4…140.7 °C
MDL Number: MFCD01862362
CAS Number: 367453-05-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adff553c3a22c98df6b14f46760b01d5ae9e1596&vid=0e1658c8dfa2b0a719bb2660ca5a9b95346a1870&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9511a8050ec2cab29531f22403b486b11fb46abc&vid=8b71f7198b5f8e5eab377ebd07f244eefcfe5c83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR