H-Hyp(tBu)-OH ≥99.5%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0418251GM
100279-820EA 122.92 USD
100279-820
H-Hyp(tBu)-OH ≥99.5%, powder
H-Hyp(tBu)-OH

  • White fine flowing powder
  • Store at 0-8 °C

Formula: C₉H₁₇NO₃
MW: 187.239 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD00153445
CAS Number: 79775-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=053b92deb6f8947fbe79318a7f0b644b0587f0a1&vid=52d500be7bc1ab7340de6c349a8ef71c12bc1459&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2995ffda69b67740a95a52f0600962d705a4fb3&vid=833054a76cf65c54d0c5ad8b7f2b159cd0e9fa5e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR