4-Hydroxy-2-methoxybenzaldehyde ≥99.5%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

04180050GM
100279-790EA 411.92 USD
100279-790
4-Hydroxy-2-methoxybenzaldehyde ≥99.5%, powder
4-Hydroxy-2-methoxybenzaldehyde

  • Yellowish powder
  • Store at 0-8 °C

Formula: C₈H₈O₃
Melting Pt: 152…162 °C
MDL Number: MFCD00051964
CAS Number: 18278-34-7
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e6314841495dc9c519c83738515af5c61b00d1&vid=91e6314841495dc9c519c83738515af5c61b00d1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR