N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-Tyr-OH, N-Fmoc-L-tyrosine

0414131GM
100279-194EA 34 USD
100279-194
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥98%, powder
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine

  • White to Off-White powder
  • Store at 0-8 °C

Formula: C₂₄H₂₁NO₅
MW: 403.43 g/mol
Melting Pt: 180…185 °C
MDL Number: MFCD00134890
CAS Number: 92954-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cfe8959c01699b0a0839a6d0f714d979d55bd78&vid=0cfe8959c01699b0a0839a6d0f714d979d55bd78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR