Fmoc-L-Tyr(Me)-OH ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

0414101GM
100279-190EA 170 USD
100279-190
Fmoc-L-Tyr(Me)-OH ≥99%, crystalline powder
Fmoc-L-Tyr(Me)-OH

  • White fine crystalline powder
  • Store at 0-8 °C

Formula: C₂₅H₂₃NO₅
MW: 417.47 g/mol
Melting Pt: 160.5…162 °C
MDL Number: MFCD00153368
CAS Number: 77128-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a80f3f035278e3b66626e4298e36584e7744938&vid=9a80f3f035278e3b66626e4298e36584e7744938&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR