Fmoc-D-Thr(tBu)-OH ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine, Fmoc-D-Thr(tBu)-OH, (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-D-threonine

0413881GM
100279-154EA 167.38 USD
100279-154
Fmoc-D-Thr(tBu)-OH ≥99%, powder
Fmoc-D-Thr(tBu)-OH

  • White powder
  • Store at 0-8 °C

Formula: C₂₃H₂₇NO₅
MW: 397.471 g/mol
Melting Pt: 128.5…137.3 °C
MDL Number: MFCD00080292
CAS Number: 138797-71-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d9a1b1101195da7a8841f9403a43f34de4e35eb&vid=ed41f344ba7d6de62a193c1fb293809679c985d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR