Fmoc-Glu-OtBu ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu

0412721GM
100278-976EA 154.31 USD
100278-976
Fmoc-Glu-OtBu ≥99%
Fmoc-Glu-OtBu
Formula: C₂₄H₂₇NO₆
MW: 425.47 g/mol
MDL Number: MFCD00065648
CAS Number: 84793-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90fa26ca319066c4d3ba0dfa9db0b5252a9412c5&vid=90fa26ca319066c4d3ba0dfa9db0b5252a9412c5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a378974394bdfd78137f280646a8bad00902a1f&vid=3a378974394bdfd78137f280646a8bad00902a1f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR