Oleyl palmitate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

451638100MG
100278-342EA 255 USD
100278-342
Oleyl palmitate ≥99%
Oleyl palmitate
MDL Number: MFCD00056327
CAS Number: 2906-55-0
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5593db3b6359eefc782b5519518a4bfd0f7f71ef&vid=5593db3b6359eefc782b5519518a4bfd0f7f71ef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR