Hexadecyl palmitate ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate

451616100MG
100278-338EA 255 USD
100278-338
Hexadecyl palmitate ≥99%, powder
Hexadecyl palmitate

  • Powder
  • Store in a cool, dry place

Formula: C₃₂H₆₄O₂
MW: 480.86 g/mol
Melting Pt: 55…56 °C
MDL Number: MFCD00053739
CAS Number: 540-10-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=781b17298a4ab853414aa9cd949021faf3d1892e&vid=781b17298a4ab853414aa9cd949021faf3d1892e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR