Myristyl myristate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tetradecyl tetradecanoate

451414100MG
100278-324EA 251.08 USD
100278-324
Myristyl myristate ≥99%
Myristyl myristate
Formula: C₂₈H₅₆O₂
MW: 424.7431 g/mol
MDL Number: MFCD00056201
CAS Number: 3234-85-3
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a799529d3fac32827b51b79c42f884a388e7667&vid=0a799529d3fac32827b51b79c42f884a388e7667&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR