1,3-Dilinolein ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Hydroxypropane-1,3-diyl (9Z,9'Z,12Z,12'Z)-bis(octadeca-9,12-dienoate)

32182325MG
100278-286EA 231.46 USD
100278-286
1,3-Dilinolein ≥99%
1,3-Dilinolein
Formula: C₃₉H₆₈O₅
MW: 616.954 g/mol
MDL Number: MFCD00056310
CAS Number: 15818-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef5bff83fec118be89a94d607e09f87495bb299&vid=6ef5bff83fec118be89a94d607e09f87495bb299&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR