1,3-Diolein ≥99%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glyceryl dioleate

32181325MG
100278-282EA 251.08 USD
100278-282
1,3-Diolein ≥99%, liquid
1,3-Diolein
Formula: C₃₉H₇₂O₅
MW: 621 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042913
CAS Number: 2465-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2abcdcb2b4e818ea5d31ee60ab772bfe26a90562&vid=2abcdcb2b4e818ea5d31ee60ab772bfe26a90562&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR