1,3-Dielaidin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Glycerol-1,2-and-1,3-dioleate

321810100MG
100278-280EA 251.08 USD
100278-280
1,3-Dielaidin ≥99%
1,3-Dielaidin
Formula: C₃₉H₇₂O₅
MW: 621 g/mol
MDL Number: MFCD00042913
CAS Number: 25637-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR