1-Monolinoleoyl-rac-glycerol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Monolinolein (9C,12C)

31182025MG
100278-274EA 219.69 USD
100278-274
1-Monolinoleoyl-rac-glycerol ≥99%
1-Monolinoleoyl-rac-glycerol

Store in a cool, dry place

Formula: C₂₁H₃₈O₄
MW: 354.531 g/mol
MDL Number: MFCD00056654
CAS Number: 2277-28-3
UN: 12352100

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR