Monoolein ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 9-Octadecenoicacid(Z)-,2,3-dihydroxypropylester, 1-(cis-9-Octadecenoyl)-rac-glycerol, Glyceryl cis-9-octadecenate hygroscopic, DL-α-Monoolein, rac-Glycerol 1-monooleate, 1-Oleoyl-rac-glycerol, 1-Mono(cis-9-octadecenoyl)-rac-glycerol, 1-Monleoyl-rac-glycerol, Glyceryl monooleate

311810100MG
100278-272EA 317.77 USD
100278-272
Monoolein ≥99%
Monoolein

  • "ERROR:#N/A"

Formula: C₂₁H₄₀O₄
MW: 356.547 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042735
CAS Number: 111-03-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace7f78e7a2c68983996f3fa3129d15f80a4f4c7&vid=f3f18c51fa0b70c92418e9494215485535c1e2f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR