3,5-Difluorobenzhydrazide ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 3,5-Difluorobenzohydrazide, 3,5-Difluorobenzoic hydrazide, 3,5-Difluorobenzene-1-carbohydrazide, 3,5-Difluorobenzoic acid hydrazide

266481GM
100278-204EA 111.15 USD
100278-204
3,5-Difluorobenzhydrazide ≥97%
3,5-Difluorobenzhydrazide
Formula: C₇H₆F₂N₂O
MW: 172.13
Melting Pt: 168…170 °C
MDL Number: MFCD01310810
CAS Number: 244022-63-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1d5bdb489c17970bf27d537087bee574173e816&vid=c1d5bdb489c17970bf27d537087bee574173e816&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR