11(Z),14(Z)-Eicosadienoic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

102002100MG
100277-870EA 320.38 USD
100277-870
11(Z),14(Z)-Eicosadienoic acid ≥99%
11(Z),14(Z)-Eicosadienoic acid
MDL Number: MFCD00673431
CAS Number: 2091-39-6
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4c0b39d565d57319a7a4414602cb126ef20a82b&vid=a4c0b39d565d57319a7a4414602cb126ef20a82b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR