2-Chloro-5-(trifluoromethyl)benzylamine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

0932745GM
100277-738EA 137.31 USD
100277-738
2-Chloro-5-(trifluoromethyl)benzylamine ≥98%
2-Chloro-5-(trifluoromethyl)benzylamine
Formula: C₈H₇ClF₃N
MW: 209.6
Density: 1.33
MDL Number: MFCD00190123
CAS Number: 15996-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4f19a3e1a3aa867f4a53a29afe4d3c6368b7053&vid=a4f19a3e1a3aa867f4a53a29afe4d3c6368b7053&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR