n-Dotriacontane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Dotriacontane

053200100MG
100277-534EA 258.92 USD
100277-534
n-Dotriacontane ≥99%
n-Dotriacontane

  • Solid
  • Store in a cool, dry place

Formula: H₃C(CH₂)₃₀CH₃
MDL Number: MFCD00009411
CAS Number: 544-85-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e65d7c309e8288ead11f47478845326adc444e1c&vid=e65d7c309e8288ead11f47478845326adc444e1c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR