n-Octacosane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-octacosane

052800100MG
100277-530EA 255 USD
100277-530
n-Octacosane ≥99%
n-Octacosane
Formula: H₃C(CH₂)₂₆CH₃
MW: 394.77 g/mol
Boiling Pt: 432
MDL Number: MFCD00009355
CAS Number: 630-02-4
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a44026c1c95995efa7de9b8305b60ae37741f57&vid=9a44026c1c95995efa7de9b8305b60ae37741f57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR