n-Hexacosane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

052600100MG
100277-528EA 255 USD
100277-528
n-Hexacosane ≥99%
n-Hexacosane
Formula: H₃C(CH₂)₂₄CH₃
MDL Number: MFCD00009354
CAS Number: 630-01-3
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=256c021bd70a6c9714e9ae1d62ca701ed743c61e&vid=256c021bd70a6c9714e9ae1d62ca701ed743c61e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR