Boc-L-Nle(6-OH) ≥99.5%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-6-OH-Nle-OH, (S)-2-((tert-Butoxycarbonyl)amino)-6-hydroxyhexanoic acid

047341GM
100277-366EA 326.92 USD
100277-366
Boc-L-Nle(6-OH) ≥99.5%, crystalline powder
Boc-L-Nle(6-OH)

  • White crystalline powder
  • Store at 0-8 °C

Formula: C₁₁H₂₁NO₅
MW: 247.3 g/mol
Melting Pt: 106…114 °C
MDL Number: MFCD01862292
CAS Number: 77611-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=953e501ecf539d745b64bb23f39d0634250ecf3d&vid=953e501ecf539d745b64bb23f39d0634250ecf3d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR