Boc-D-Dap(Aloc)-OH DCHA

Supplier: INDOFINE CHEMICAL COMPANY MS

046331GM
100277-302EA 328.23 USD
100277-302
Boc-D-Dap(Aloc)-OH DCHA
Boc-D-Dap(Aloc)-OH DCHA
Formula: C₁₂H₂₁N₂O₆·C₁₂H₂₃N
MW: 469.56 g/mol
MDL Number: MFCD00800347
CAS Number: 204197-26-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd7195681efe5abda7e5d5361eeaa05e29efa058&vid=c64b782d4d833975c08fb63b90efe5363dc10bf1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR