N-Benzylglycine hydrochloride ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Bzl-Gly-OH·HCl, Bzl-Gly-OH.HCl

044455GM
100277-172EA 102 USD
100277-172
N-Benzylglycine hydrochloride ≥99%, powder
N-Benzylglycine hydrochloride

  • White powder
  • Store at 0-8 °C

Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
MDL Number: MFCD00156934
CAS Number: 7689-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR