1-Aminocyclopropanecarboxylic acid ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-Amino-1-cyclopropanecarboxylic acid

042645GM
100277-022EA 122.92 USD
100277-022
1-Aminocyclopropanecarboxylic acid ≥99%, powder
1-Aminocyclopropanecarboxylic acid

  • White powder
  • Store at 0-8 °C

Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00009944
CAS Number: 22059-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9afe2ab41a088e542b93ef19831d8db63308783&vid=e9afe2ab41a088e542b93ef19831d8db63308783&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR