L-Tyrosine benzyl ester ≥99%, crystalline powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Tyr-OBzl, Benzyl tyrosinate, (S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid benzyl ester

04262025GM
100277-006EA 209.23 USD
100277-006
L-Tyrosine benzyl ester ≥99%, crystalline powder
L-Tyrosine benzyl ester

  • White crystalline powder
  • Store in a cool, dry place

Formula: C₁₆H₁₇NO₃
MW: 271.316 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00191034
CAS Number: 42406-77-9
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2122b12cd25f1b06828492bbda1045de89944ec6&vid=2122b12cd25f1b06828492bbda1045de89944ec6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR