Tri-O-acetyl-D-glucal ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (-)-Tri-O-acetyl-D-glucal, 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol 3,4,6-Tri-O-acetyl Ether, (2R,3S,4R)-2-(Acetoxymethyl)-3,4-dihydro-2H-pyran-3,4-diyl diacetate

04253825GM
100276-944EA 146.46 USD
100276-944
Tri-O-acetyl-D-glucal ≥99%, powder
Tri-O-acetyl-D-glucal

  • White to off-white powder
  • Store in a cool, dry place

Formula: C₁₂H₁₆O₇
MW: 272.25 g/mol
Melting Pt: 49…57 °C
MDL Number: MFCD00063253
CAS Number: 2873-29-2
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=201a8676fc991160a6b6730c39dc9f2fa4bef219&vid=90dd49e30c5d7a0fdb73b469a8528ae133f661c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR