(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic Acid, H-D-Tic-OH, D-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

0424771GM
100276-886EA 37.92 USD
100276-886
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥99%
(R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

  • White yellow solid
  • Store at 0-8 °C

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00144038
CAS Number: 103733-65-9
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee6240c346d431f8ab24ee26d850c84c569ff1c1&vid=ee6240c346d431f8ab24ee26d850c84c569ff1c1&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba9e254aba0263f0b4743c68e9448f4899bae043&vid=ba9e254aba0263f0b4743c68e9448f4899bae043&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR