Cbz-D-Phe-L-Pro

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Z-D-Phe-Pro-OH

0422671GM
100276-712EA 253.69 USD
100276-712
Cbz-D-Phe-L-Pro
Cbz-D-Phe-L-Pro
Formula: C₂₂H₂₄N₂O₅
MW: 396.443 g/mol
MDL Number: MFCD00238427
CAS Number: 17460-56-9
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2271e9e91fd95a149071c4375127dfa6c35f94ca&vid=2271e9e91fd95a149071c4375127dfa6c35f94ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR