Fmoc-D-4-bromophenylalanine ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Fmoc-4-bromo-D-Phe-OH, 4-Bromo-N-Fmoc-D-phenylalanine, Fmoc-D-Phe(4-Br)-OH

0413521GM
100276-110EA 143.85 USD
100276-110
Fmoc-D-4-bromophenylalanine ≥99%, powder
Fmoc-D-4-bromophenylalanine

  • White powder
  • Store at 0-8 °C

Formula: C₂₄H₂₀BrNO₄
MW: 466.331 g/mol
MDL Number: MFCD00273459
CAS Number: 198545-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b2d3446a53022690c08fe91223aa7acc1b8ad15&vid=9b2d3446a53022690c08fe91223aa7acc1b8ad15&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR