Methyl-2-hydroxyeicosanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

24200250MG
100275-020EA 583.23 USD
100275-020
Methyl-2-hydroxyeicosanoate ≥98%
Methyl-2-hydroxyeicosanoate

Store in a cool, dry place

Formula: C₂₁H₄₂O₃
MW: 342.564 g/mol
MDL Number: MFCD00083368
CAS Number: 16742-49-7
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3537bf8634ea3fe97cea6bb8a5b066fe35de527f&vid=3537bf8634ea3fe97cea6bb8a5b066fe35de527f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR