Methyl-17-methyloctadecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21181725MG
100274-950EA 623.77 USD
100274-950
Methyl-17-methyloctadecanoate ≥98%
Methyl-17-methyloctadecanoate
MDL Number: MFCD00083425
CAS Number: 55124-97-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7183c1ede9756aae61026889b0e9b4bfd1591aa2&vid=7183c1ede9756aae61026889b0e9b4bfd1591aa2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR