Methyl-12-methyltetradecanoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

21141225MG
100274-924EA 562.31 USD
100274-924
Methyl-12-methyltetradecanoate ≥98%
Methyl-12-methyltetradecanoate
MW: 256.43 g/mol
MDL Number: MFCD00053774
CAS Number: 5129-66-8
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b7257cf4403b0848da4c4526a2c549c9927e6a&vid=b3b7257cf4403b0848da4c4526a2c549c9927e6a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR